Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z606340

Eppendorf® microtubes 3810X

volume 1.5 mL, colorless

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702

material

conical bottom tube
polypropylene

sterility

non-sterile

feature

RCF 25,000 × g
lid

packaging

case of 1000

manufacturer/tradename

Eppendorf® 0030 125.150
Eppendorf® 022364111

availability

not available in USA and Canada

volume

1.5 mL

color

colorless

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katayi Mwila Kazimbaya et al.
PloS one, 15(11), e0240714-e0240714 (2020-11-11)
Despite contributing to a significant reduction in rotavirus associated diarrhoea in highly burdened low- and middle-income countries, live attenuated, oral rotavirus vaccines have lower immunogenicity and efficacy in these settings in comparison to more developed countries. Breastmilk has been implicated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service