Key Documents
Z606332
Eppendorf® PCR tubes
capacity 0.5 mL
About This Item
UNSPSC Code:
41103037
NACRES:
NB.11
Pricing and availability is not currently available.
material
cap (hinged lid clip)
colorless tube
conical bottom tube
polypropylene
feature
RNase and DNase free
packaging
pkg of 500 tubes (bulk packed)
manufacturer/tradename
Eppendorf® 0030 124.502
availability
not available in USA and Canada
technique(s)
PCR: suitable
capacity
0.5 mL
General description
Thin-wall PCR tubes with a contamination shield at the front edge of the lid to prevent accidental contact with the inner surface when the tube is opened.
Matt writing surface on lid
Chem/Phys Resistance
Autoclavable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)