Key Documents
Z606308
Eppendorf® Safe-Lock microtubes, PCR clean
capacity 0.5 mL
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
Pricing and availability is not currently available.
material
cap (hinged lid clip)
colorless
conical bottom tube
polypropylene
feature
RCF 30,000 × g
RNase and DNase free
packaging
case of 500
manufacturer/tradename
Eppendorf® 0030 123.301
availability
not available in USA and Canada
technique(s)
PCR: suitable
capacity
0.5 mL
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)