Key Documents
Z605050
Sigma-Aldrich
Eppendorf® UVette®
pack of 80 disposable cuvettes, spectral range 220-1,600 nm
Synonym(s):
biophotometer, cuvette uv, uv cuvette
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
material
plastic
description
UV-Vis transparent between 220 and 1600 nm
individual blister packs to prevent contamination and scratches
feature
pack of 80 disposable cuvettes
manufacturer/tradename
Eppendorf® 0030 106.300
parameter
220-1,600 nm spectral range
pathlength
10x2 mm
absorption
≤0.5 at 260 nm (absorbance units)
foreign activity
DNA, Rnase and protein free
General description
Two optical path lengths with a single rotation! Samples with a normal concentration can be measured using the 10 mm optical path length. For samples with a higher concentration, simply rotate the UVette by 90° and use the shorter 2 mm optical path length. 4 optical windows, crystal clear, UV and VIS transparent in the range 200-1600 nm. Funnel-shaped base prevents capillary effect, ensures sample in center of measuring area. The new UVette is ideal for use with the Eppendorf BioPhotometer. In other photometers or spectrometers the Uvette is inserted with the aid of an adapter. This adapter aligns the UVette to the corresponding height of the optical light path and also serves as an aperture for optimal light guidance and use of the small measuring volume. Individually packaged, UVettes are DNA, RNase, and protein-free, and are protected from scratches and dust.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
UVette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)