Key Documents
Z601551
Hettich® centrifuge buckets
Bio-containment lid for bucket/carrier 1381
About This Item
UNSPSC Code:
41103910
Pricing and availability is not currently available.
manufacturer/tradename
Hettich 1382
availability
available only in EU
not available in USA and Canada
technique(s)
centrifugation: suitable
General description
  • Bio-containment lid for carrier 1381 (Z601543)
  • Bucket/Carrier 1381 (Z601543) is compatibile with Swing-out rotors 1324, 1494, 1624; 1724
  • Swing-out Rotors (1324, 1494, 1624; 1724) are compatible with the following Hettich Centrifuges: ROTOFIX 32A; ROTINA 38/38R; UNIVERSAL 320/320R
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)