Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z4525

Sigma-Aldrich

Anti-ZIC2 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 55 kDa

species reactivity

rodent, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZIC1(7545)
mouse ... Zic2(22772)
rat ... Zic2(361096)

Immunogen

synthetic peptide corresponding to human ZIC2 (amino acids 162-176) conjugated to BSA.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohe Chen et al.
Molecular medicine reports, 19(1), 508-514 (2018-11-30)
MicroRNAs (miRNAs) have been demonstrated to regulate the progression of numerous types of cancer, including acute myeloid leukemia (AML). Previous studies demonstrated that miR‑1271‑5p functions as a tumor suppressor; however, the roles of miR‑1271‑5p in AML remain unknown. In the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service