Key Documents
Z381489
Heating blocks for Techne® Dri-Block® heater
holds 30, 0.5 mL microcentrifuge tubes
About This Item
UNSPSC Code:
41102404
feature
Holds 30 x 0.5 mL tubes
manufacturer/tradename
Techne F4465
General description
Interchangeable blocks machined of aluminum for rapid heat transfer. Close tolerances assure efficient, rapid heating. Each block has a threaded thermometer hole for independent temperature monitoring, which also fits the block extraction tool.
Legal Information
Dri-Block is a registered trademark of Techne, Inc.
Techne is a registered trademark of Techne, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)