Type I collagen is an excellent adhesion factor for many cell types, including epithelial, endothelial, muscle, and neuronal lines. CollagenPRO COAT is highly purified and processed to offer the least immunogenic collagen-coated cultureware currently available. The processing also improves clarity (the coated surfaces are more transparent) and provides a natural environment for attachment and rapid proliferation. All vessels coated with CollagenPRO COAT have flat bottoms.
Application
A T-25 configuration for the attachment of epithelial, endothelial, muscle, and neuronal cell types.
Legal Information
CollagenPRO is a trademark of Novagen, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)