Key Documents
Z380512
Sigma-Aldrich
CollagenPRO COAT cultureware
culture dish, 35 mm
About This Item
UNSPSC Code:
41100000
General description
Type I collagen is an excellent adhesion factor for many cell types, including epithelial, endothelial, muscle, and neuronal lines. CollagenPRO COAT is highly purified and processed to offer the least immunogenic collagen-coated cultureware currently available. The processing also improves clarity (the coated surfaces are more transparent) and provides a natural environment for attachment and rapid proliferation.
All vessels coated with CollagenPRO COAT have flat bottoms.
Application
A 35 nm configuration for the attachment of epithelial, endothelial, muscle, and neuronal cell types.
Legal Information
CollagenPRO is a trademark of Novagen, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)