Key Documents
Z379387
Hanna pHel pH meter
Model pHel
About This Item
UNSPSC Code:
41115603
material
plastic body
feature
calibration manual two-point
manufacturer/tradename
Hanna
pH measuring range
0-14, resolution: 0.1
size
200 mm × 28 mm × 20 mm
weight
46 g
battery power
2 × 1.4 V button cell, 3000 hr life (continuous operation)
General description
Double-junction gel-filled electrode
The convenience of a pocket meter with the reliability of a professional double-junction electrode. The long, slim stem makes it possible to measure small samples in small vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)