Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z379301

Sigma-Aldrich

Side Loader collar for Deepwell plate

Synonym(s):

Deepwell 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
size:
96 wells
sterility:
non-sterile

sterility

non-sterile

manufacturer/tradename

Bel-Art F37860-0005

size

96 wells

color

none

Looking for similar products? Visit Product Comparison Guide

General description

  • Side loader collars are required for side loading Beckman BioMek systems.
These high-form polypropylene 96 well plates expand the flexibility of multichannel or automated pipetting systems. Use for non-binding protein assays, enzyme or receptor assays, or with cell harvesters. The 2 mL per well capacity makes it possible to do serial dilution procedures that are impossible with normal multiwell plates.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service