Key Documents
Z379069
CoverWell perfusion chambers
1 chamber per slide, round chamber, volume 750 μL
Synonym(s):
imaging chamber, in situ hybridization chamber
About This Item
UNSPSC Code:
41100000
material
round chamber
manufacturer/tradename
Grace Bio-Labs
D × diam.
2.5 mm × 20 mm
volume
750 μL
General description
CoverWell perfusion chambers are press-to-seal covers which form multiple cell incubation or cytochemistry chambers when pressed to glass coverslips or microscope slides. Reagents can be quickly added or removed through dual access ports without disturbing or cross-contaminating specimens. Suitable for live cell imaging on upright or inverted, visible light and fluorescence microscopes.
Legal Information
CoverWell is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)