Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z378666

Sigma-Aldrich

Sigma-Solohill microcarrier beads

ProNectin F coated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

material

polystyrene beads

bead size

125-212 μm

particle size

125-212 μm

density

1.02 g/cm3 (lit.)

Looking for similar products? Visit Product Comparison Guide

General description

ProNectin F is a recombinant non-animal-source polymer that promotes attachment of even weakly adherent cell lines. Optimized for pharmaceutical applications, this bead performs well in serum-free and even in protein-free media. The coating consists of multiple copies of the human fibronectin RGD attachment domain. Beads can be autoclaved in deionized water or PBS.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan Chen et al.
Cell and tissue research, 359(2), 577-587 (2014-11-02)
Tendon calcification is common in the Achilles tendon, and injuries affect not only athletes, but also the general population. However, the underlying cellular mechanisms are not yet fully understood. In this study, we isolated healthy human tenocytes and subjected them

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service