Z377813
ABI DNA Sequencing Unit Accessories
Sharktooth Comb, thickness 0.2 mm × 36 wells
About This Item
thickness
0.2 mm × 36 wells
General description
Special features of Step Plates:
- Front plate is beveled to create a thicker gel in the comb area, giving enough sample capacity for 96 lanes
- Gel casting is standard, but uses a 0.4 mm casting comb and sharktooth comb
Special features of Microtrough 96 plates:
- Microtroughs notched inside the front plate provide enough sample capacity for 96 lanes
- A special Mylar plug instead of a standard casting comb
- Microtroughs provide greater loading area and guide your pipette tops for more accurate sample placement, eliminating the need for expensive loaders or specialized tips
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)