Key Documents
Z377511
Sigma-Aldrich
Scienceware® autoradiography film binder
Binder
About This Item
UNSPSC Code:
41100000
General description
Includes 10 sheet protectors
Sturdy, vinyl-covered, 10-ring loose-leaf binder for storing and protecting autoradiography films. Supplied with 10 pre-punched protectors, each of which can accommodate two 14 × 17 inch (35 × 43 cm) films. With additional protectors, capacity of binder is about 40 films.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)