Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z377422

Nalgene® FastCap bottle-top filter unit

pore size 0.2 μm

Synonym(s):

bottle top filter, nalgene filter unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
NACRES:
NB.24

material

polyethersulfone membrane

sterility

sterile

feature

cap plug seal

manufacturer/tradename

Nalgene® 298-9020

filter capacity

5000 mL

filter diam.

90 mm

pore size

0.2 μm

fitting

bottle neck (Fits up to 53 mm)

Looking for similar products? Visit Product Comparison Guide

General description

Filter from the prep flask directly into the receiver. These bottle-top filters with low protein binding PES membrane have a universal FastCap fitting. Just place it on the mouth of the receiver and apply vacuum. Filters up to 5 liters. Sterilized by γ-irradiation.

Legal Information

FastCap is a trademark of Thermo Fisher Scientific or its subsidiaries
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Characterize mAb monomers, aggregates, and fragments using SEC-UV workflow with Zenix® and Zenix®-C SEC columns.

Protocols

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service