Key Documents
Z376027
Genesys 20 spectrophotometer
With U.S. cord
Synonym(s):
spectroscopy uv visible, ultraviolet spectrophotometer, uv vis spectroscopy
About This Item
UNSPSC Code:
12352200
General description
Features a sleek, new design with up-to-date electronics and the latest optical components. Measures absorbance or transmittance over a range of 325-1100 nm and can be programmed to print out concentrations. Uses 10 mm square cuvettes or 10 mm test tubes. Includes single-position cell holder, 5 disposable cuvettes, RS-232C interface, Centronics-compatible printer port, dust cover, and owner′s manual. 100-240 V compatible.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Zhi-Cheng Xu et al.
Molecular medicine reports, 15(1), 417-422 (2016-12-14)
Hair follicle stem cells (HFSCs) are increasingly used as a stem cell paradigm in vascular tissue engineering due to the fact that they are a rich source of easily accessible multipotent adult stem cells. Promising results have been demonstrated with