Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z374466

Sigma-Aldrich

Scienceware® cloning discs

diam. 6.4 mm ( 1/4 in.)

Synonym(s):

cloning disc, cloning ring, cloning rings

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106618
NACRES:
NB.22

sterility

sterile; γ-irradiated

manufacturer/tradename

Bel-Art

diam.

6.4 mm ( 1/4 in.)

Looking for similar products? Visit Product Comparison Guide

General description

Paper discs transfer cells quickly and easily to a 24 well plate for growth. Discs eliminate leak problems associated with cloning cylinders. Risk of contamination and drying out is reduced because cells are exposed only a fraction of the time it would take to apply a cloning cylinder.
Contains 2 vials of 50 cloning discs.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service