Key Documents
Z374261
Sigma-Aldrich
Intensifying screens BioMax® Transcreen®
LE for 14 in. × 17 in. film
Synonym(s):
autoradiography cassettes, xray film cassettes
About This Item
UNSPSC Code:
41105341
NACRES:
NB.31
General description
A new concept in intensifying screens provides improved resolution and maximum sensitivity and can now enhance exposures with low-energy β-emitters.
Conventional intensifying screens capture particles after they have passed through the film and "reflect" them back as light. Low-energy particles can never reach a conventional intensifying screen. TranScreens, placed between the gel or blot and the film, intercept particles and "transmit" them to the film as visible light. Choose from:
  • LE for low-energy β-emitters like 3H, 14C, 33P, 35S, 45Ca, and 59Fe
  • HE for high-energy β-emitters like 32P and γ-emitters like 125I
Both are optimized for use with BioMax MS film, but can be used with other films.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Transcreen is a registered trademark of Kodak
Choose from one of the most recent versions:
Certificates of Analysis (COA)