Key Documents
Z373036
Sigma-Aldrich
Plate separator wedges
provides separation of gel plates after electrophoresis, pkg of 2 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
polycarbonate
packaging
pkg of 2 ea
General description
Small polycarbonate wedges that facilitate safe separation of gel plates after electrophoresis.
Just insert a wedge between the plates − either just inside the "ear" at the top or just inside the spacer at the bottom − and push with steady force. A slight twist sometimes helps. Can also be used to scrape gel residue off without scratching the glass.
Choose from one of the most recent versions:
Certificates of Analysis (COA)