Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z372366

Sigma-Aldrich

Horizontal submarine electrophoresis units

gel size 7.5 cm × 10 cm, base unit, dual mode, mini-gel 7.5 cm wide, pink

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

material

acrylic

grade

electrophoresis grade

gel size

7.5 cm × 10 cm

color

pink

Looking for similar products? Visit Product Comparison Guide

General description

Submarine gels may be formed directly on the UV-transparent gel bed using small sliding baffles to contain the agarose as it cools, or outside the unit using a UV-transparent gel tray sealed with tape. Gel is 7.5 cm wide × 10 cm long. Different colors are offered for any color-coding your lab requires (e.g., RNA only).
Combs and tape are not included. Gel tray is not included with the Mini-Gel unit.
Base unit, safety cover, power leads, and all other necessary parts are included.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service