Submarine gels may be formed directly on the UV-transparent gel bed using small sliding baffles to contain the agarose as it cools, or outside the unit using a UV-transparent gel tray sealed with tape. Gel is 7.5 cm wide × 10 cm long. Different colors are offered for any color-coding your lab requires (e.g., RNA only). Combs and tape are not included. Gel tray is not included with the Mini-Gel unit. Base unit, safety cover, power leads, and all other necessary parts are included.
Other Notes
Also called a midi-gel, this is a double-wide mini-gel format for high-throughput sample screening.
Choose from one of the most recent versions:
Certificates of Analysis (COA)