Key Documents
Z369861
Diagnostic Molecular Microbiology: Principles and Applications
About This Item
ISBN-10:
1-55581-056-X
ISBN-13:
978-1-55581-056-6
UNSPSC Code:
55101509
publication info
D.H. Pershing, ed., American Society of Microbiology, 1993, 664 pp., soft cover
manufacturer/tradename
American Society Of Microbiology
General description
DNA techniques in the microbiology laboratory comprehensively reviewed, with four editors contributing their particular expertise and viewpoints. Cutting edge techniques like nucleic acid probes and in vitro amplification, and concepts of quality control in PCR processes are applied to identification of pathogens, known and novel in over 60 detailed protocols.
Choose from one of the most recent versions:
Certificates of Analysis (COA)