Key Documents
Z368776
Finnpipette® digital single channel pipette
5-50 μL, resolution: 0.5 μL
About This Item
UNSPSC Code:
41121500
feature
electronic: no
single-channel
manufacturer/tradename
Finn
measuring range
5-50 μL, resolution: 0.5 μL
volume
50 μL
General description
New ergonomic design with all the best features of earlier Finnpipette lines: fully autoclavable without disassembly; certified factory calibration and easy in-lab recalibration (tool and instructions included); soft-touch tip ejection provides more ejector force with less thumb pressure.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)