Z368199
Eppendorf® centrifugal vacuum concentrator
Complete Model 5301, AC/DC input 115 V AC, 50 - 60 Hz
All Photos(1)
About This Item
UNSPSC Code:
41103900
manufacturer/tradename
Eppendorf®
availability
not available in EU
General description
Complete system includes solvent-resistant PTFE vacuum pump, vapor trap, and rotor for 48 × 1.5-2 mL tubes
Concentrates aqueous and alcoholic solutions quickly and cleanly right in their microtube containers. The unit applies gentle heat and vacuum to evaporate the solvent, while centrifugation prevents bumping and foaming. Choice of four temperature settings (ambient, 30, 45, and 60 °C) allows processing of a variety of samples. Vacuum is automatically released before the centrifuge turns off. The 48-place rotor for 1.5-2 mL tubes and the 72-place rotor for 0.5 mL tubes can be stacked two high for double the throughput.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)