Contains three accessory platforms for the Hi-Lo Rocker.
Kit includes all hardware necessary to add 3 accessory platforms to the Hi-Lo Rocker for a total of 4 platforms. Made of stainless steel for years of dependable service. Platforms can also be autoclaved to prevent contamination of incubators.
Made in the USA
Features and Benefits
Contains Three stainless steel platforms
All hardware included.
Transforms Hi-Lo Rocker into a Four platform unit.
Other Notes
Note: platform accessory for Z367745; Z367753
Choose from one of the most recent versions:
Certificates of Analysis (COA)