Key Documents
Z367761
Hi/Lo profile rocker accessories
one add-on platform assembly
About This Item
UNSPSC Code:
41100000
NACRES:
NB.73
manufacturer/tradename
IBI ACCRC00P1
General description
Contains one accessory platform for the Hi-Lo Rocker.
Kit includes all hardware necessary to add 1 accessory platform to the Hi-Lo Rocker for a total of 2 platforms. Made of stainless steel for years of dependable service. Platforms can also be autoclaved to prevent contamination of incubators.
Made in the USA
Features and Benefits
  • Contains One stainless steel platform
  • All hardware included.
  • Transforms Hi-Lo Rocker into a Two platform unit.
Other Notes
Note: platform accessory for Z367745; Z367753
Choose from one of the most recent versions:
Certificates of Analysis (COA)