Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z365556

Supelco

Preval spray unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

technique(s)

thin layer chromatography (TLC): suitable

General description

Compact, well-designed spray canister delivers fine mist with CFC-free propellant. Permits control of reagent delivery, for even coverage and minimal waste. Gas canister serves as handle; cap fits standard 38-400 thread reagent bottles. Kit includes one propellant canister and one 6-ounce glass bottle.

Pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Gas 1A

Storage Class

2B - Aerosol cans and lighters

wgk_germany

WGK 1

flash_point_f

-155.2 °F - closed cup

flash_point_c

-104 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service