Prevents cross-contamination due to over-pipetting. Incorporates a dual hydrophilic-hydrophobic 0.8 μm filter that prevents further use once wet. Includes four filters. Order extra filters separately.
Safe, convenient instruments for converting pipettes into hand-operated dispensers. Especially useful in confined areas such as laminar flow hoods. Can be used with glass or plastic pipettes ranging in size from 1 to 100 mL.
Legal Information
Pipet-Aid is a registered trademark of Drummond Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)