Key Documents
Z363529
Sigma-Aldrich
Scienceware® autoradiography pen
High energy level
About This Item
UNSPSC Code:
41105341
NACRES:
NB.31
General description
Higher signal emission strength than Z363510 and a slightly longer duration of signal emission time. Best used for marking x-ray films similar to Kodak XLS. It also works well when film response is lowered as with chilled films which often require intensifier screen techniques. The mark emits no Beta or other isotope emission, only visible light. Therefore, the use of an intensifier screen will not occur as a variable in the x-ray marking procedures.
Non-radioactive markers for annotating autoradiograms.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)