Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z363456

Sigma-Aldrich

Desiccator cartridges

diam. 100 mm

Synonym(s):

desiccant cartridge, desiccator, moisture sensitive protection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
24141516

packaging

pack of 6 ea

manufacturer/tradename

Bel-Art F42048-0100

diam.

100 mm

Looking for similar products? Visit Product Comparison Guide

General description

The desiccant cartridges can be regenerated by gentle overnight heating in a 50°C (122°F) oven (higher temperature can melt dish). For quicker regeneration, cut open the membrane and transport beads to a glass or porcelain dish and heat in a 150°C (302°F) oven for one hour. Silica gel is non-toxic, non-corrosive and inert. Clear polystyrene dish contains indicating silica gel and allows instant visual inspection. Porous membrane cover provides good air exchange for efficient moisture absorption.

Use to reduce moisture in desiccator cabinets or within cases containing precision laboratory instruments.

  • Silica gel beads transform to pink color when saturated with moisture
  • Transparent polystyrene dish permits instantaneous visual examination of silica gel color
  • Large or small cartridges to match requirements
  • Cartridges are shipped in individual sealed pouches, ready for immediate use.

Packaging

Cartridges are individually sealed in foil pouches.

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service