Z363448
Desiccator cartridges
diam. 65 mm
Synonym(s):
desiccant cartridge, desiccator, moisture sensitive protection
About This Item
Recommended Products
manufacturer/tradename
Bel-Art
diam.
65 mm
Looking for similar products? Visit Product Comparison Guide
General description
Use to reduce moisture in desiccator cabinets or within cases containing precision laboratory instruments.
- Silica gel beads transform to pink color when saturated with moisture
- Transparent polystyrene dish permits instantaneous visual examination of silica gel color
- Large or small cartridges to match requirements
- Cartridges are shipped in individual sealed pouches, ready for immediate use.
- Clear polypropylene dish allows instant visual inspection of silica gel, which changes in color from blue to pale pink as it absorbs moisture
- Highly porous mesh membrane covers the dish providing excellent moisture absorption
- Desiccant can be easily regenerated by heating cartridges in a 50ºC (122ºF) oven overnight or at 100ºC (212ºF) for 6 hours
- For more rapid drying, remove the membrane lid, transfer the spent gel crystals into a glass or porcelain dish, and place in a 150ºC (300ºF) oven for one hour
- Individually foil sealed
Packaging
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Carc. 1B - Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service