Key Documents
Z363162
Sigma-Aldrich
Carestream® BioMax® MS Intensifying Screens without lead blockers
For 14 × 17 in.(35 x 43 cm) film, Cassette not included
Synonym(s):
autoradiography cassettes, xray film cassettes
About This Item
UNSPSC Code:
41105341
NACRES:
NB.31
General description
Phosphor emits green light, closely matched to the spectral sensitivity of BioMax MS film; provides highest sensitivity for 32P and 125I autoradiography.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)