Key Documents
Z363006
Sigma-Aldrich
Kodak® BioMax® MS film
Synonym(s):
autoradiography film, xray film
About This Item
UNSPSC Code:
41105341
NACRES:
NB.22
size
8 in. (20 cm) × 10 in. (25 cm)
General description
Double-emulsion film specifically engineered for biotechnology applications. Provides maximum sensitivity for detection of 32P and 125I in Northern and Southern blots, reducing quantity of isotope needed.
Sheets of film with no interleaved paper
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Articles
Background and protocols describing the various methods used by molecular biologists to detect samples of protein or nucleic acids bound to membranes.
Questions
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?