Key Documents
Z360481
Utility boxes, plastic
L × W × H 127 mm × 73 mm × 175 mm
Synonym(s):
laboratory storage, plastic storage box
About This Item
UNSPSC Code:
41123300
NACRES:
NB.51
Product Z360481 is not currently sold in your country. Contact Technical Service
material
styrene-acrylonitrile (SAN)
packaging
case of 12 ea
pack of 2 ea
manufacturer/tradename
Nalgene 5700-1000
L × W × H
127 mm × 73 mm × 175 mm
General description
Practical containers for multipurpose lab storage. Glass-clear SAN body; white LDPE lid with white SAN insert. Ideal for dry materials like powders, chromatography sheets. Rounded corners for easy cleaning; stackable for compact freezer storage. Lids snap into place, but are not leakproof.
Dimensions are L × W × H
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1