Key Documents
Z359912
Millipore
Millipore® Sterivex filter unit
GP without filling bell, pore size 0.22 μm, polyethersulfone membrane
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
material
housing (Eastar)
polyethersulfone membrane
sterility
sterile
packaging
pack of 15
manufacturer/tradename
Millipore SVGP01015
pore size
0.22 μm
General description
Suitable for sterilizing and clarifying tissue culture media and other aqueous solutions in the 100 mL to 1 liter volume range. Pressure-driven, self-venting, direct-dispensing design allows filtration into any type of container without foaming. Can be used with a peristaltic pump, pressure vessel, or syringe.
Connections are female luer-lock inlet and 3.3 mm O.D. male tubing outlet or male luer-lock outlet.
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Sterivex is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)