Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Z358487
Carestream® BioMax® XAR Film
Synonym(s):
autoradiography film, xray film
About This Item
size
7 in. (18 cm) × 9.5 in. (24 cm)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
- Coated with emulsion on both sides of the clear base.
- Recommended for chemiluminescence.
- Very versatile for autoradiography with high or low energy isotopes.
- Sensitivity can be increased with intensifying screens when using low-energy isotopes.
- When handling film in a darkroom, Kodak LED Safelight Darkroom Illuminator is recommended.
- All the XAR films are the same, just packaged differently.
Footnote
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service