Key Documents
Z354465
Sigma-Aldrich
Bar clamp
for 16.5 cm wide vertical slab gel unit
About This Item
UNSPSC Code:
41100000
Features and Benefits
Replaces bulldog spring clamps for attaching the gel sandwich to the upper reservoir. Gives even compression over entire width of gel, eliminating capillary leakage between samples and reducing glass plate fractures. Clamp is included with fixed-height units, but is optional with adjustable-height units.
Choose from one of the most recent versions:
Certificates of Analysis (COA)