Z352780
Vertical electrophoresis unit, fixed height, single gel
gel W × H 16.5 cm × 22 cm
About This Item
Recommended Products
grade
electrophoresis grade
availability
not available in EU
gel W × H
16.5 cm × 22 cm
Features and Benefits
- Made from high quality acrylic with platinum electrodes
- Long lasting silicone sponge gasket on upper reservoir for leak-free operation without grease
- Safety covers with attached safety-tip power leads
- Designed for use with unique gel-casting gasket; heavy-duty casting clamps compress gasket between glass plates, sealing without grease
For Slab Gel Electrophoresis:
- Designed to use as little as 200 mL of buffer, and as little as 5 μg of sample per well
- Use with gel casting accessories which offer the ability to cast one or two gels in a single assembly
- Use in standard slab gel electrophoresis or in the second dimension of 2D electrophoresis
For Tube Gel Electrophoresis:
- Only the two fixed-height dual-gel units accept tube gel accessories
- Tubes in four different inside diameters all have the same outside diameter so they are interchangeable in the tube gel adapter
- Two-sided operation allows up to 20 tubes to be run at a time
Each unit comes with base unit, upper and lower reservoirs with safety covers and attached power leads, and two gel-mounting spring clamps.
CE APPROVED ONLY WHERE INDICATED
Slab and tube gel accessories are not included
Choose from one of the most recent versions:
Certificates of Analysis (COA)