Key Documents
Z352683
Sigma-Aldrich
Aluminum cooling plates for Sigma-Aldrich® sequencing units
For 20 × 62 cm gels
About This Item
UNSPSC Code:
41103025
NACRES:
NB.22
availability
not available in EU
Features and Benefits
Made from 1/4 in. aluminum with electrically insulating coating. Plate provides full-width cooling for 20cm or 33cm wide gels. Plate is approximately 16cm shorter than the glass plate to leave room for the upper buffer reservoir. Needed only for adjustable height sequencing units.
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)