Z351814
Eppendorf® Maxitips for Maxipettor®
Barrel and piston assemblies
All Photos(1)
About This Item
UNSPSC Code:
41121600
sterility
non-sterile
packaging
pack of 30 ea
manufacturer/tradename
Eppendorf® 0030 050.533
Eppendorf® 022291408
maximum volume
10 mL
General description
These tips allow the Maxipettor to function like an air-displacement serological pipet with integral pipetting aid. Reusable barrel/piston assembly attaches to the Maxipettor, and 170 mm Maxitip S or G attaches to the barrel.
Specially designed valve can be added to the assembly to allow dripless pipetting of liquids with high vapor pressure.
Product features of Maxitips:
Specially designed valve can be added to the assembly to allow dripless pipetting of liquids with high vapor pressure.
Product features of Maxitips:
- Maxitip P for aspirating 1 mL to 10 mL from beakers using a positive-displacement technique
- Maxitip S for aspirating 2.5 mL to 10 mL from tall, narrow-neck vessels using an air-displacement technique
- No carryover as Maxitips are exchanged
- Valve for Maxitip ensures drip-free dispensing of liquids with a high vapor pressure
- Maxitip G is graduated for dispensing accurate volumes of non-aqueous liquids
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Maxipettor is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)