Key Documents
Z351210
Eppendorf® Repeater pipette tips
About This Item
UNSPSC Code:
41121600
material
colorless tip
description
PCR inhibitor free
sterility
sterile
feature
ATP free
DNA free (Bacterial (E. Coli))
DNA free (Human)
DNase free
RNase free
barrier: no
pyrogen free
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 022495101
General description
The Combitip′s integral reservoir lets you pipet up to 48 times without refilling. Liquids contact only the reusable, disposable Combitip, eliminating cleaning and contamination of the pipet itself. Choice of Combitips gives volume selection from 1 μl to 5 mL.
The Biopur tips are sterile and certified free of DNA, RNASE, ATP, and pyrogen.
Features and Benefits
Dispenses 25, 50, 75, 100, or 125 μL
Other Notes
Browse all pipette tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)