Z351199
Eppendorf® Repeater™ pipette tips
Combitips, maximum volume 1.25 mL
About This Item
material
colorless tip
feature
barrier: no
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 0030 048.083 (European)
Eppendorf® 022261100 (USA)
maximum volume
1.25 mL
General description
The Biopur tips are sterile and certified free of DNA, RNASE, ATP, and pyrogen.
Features and Benefits
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)