Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z351067

Sigma-Aldrich

Hanna Checktemp pocket thermometer

Model HI 98501, °C display

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112303

description

without calibration certificate

manufacturer/tradename

Hanna

temperature measuring range

-50-+150 °C, accuracy: 0.3 °C, resolution: 0.1 °C

probe L × diam.

105 mm × 3 mm

weight

50 g (1.8 oz)

battery power

1.4 V button cell

Looking for similar products? Visit Product Comparison Guide

General description

Checktemp gives you the temperature read-out from -50 to 150 °C or -58 to 302 °F in seconds on a large Liquid Crystal Display with ±0.3 °C or ±0.5 °F accuracy. No breakage, No waste, No injuries, No difficulty in reading. Not even a parallax error by taking a wrong measurement due to the angle of view!

Did you ever wish that your temperature measurements could be made a little easier, in clear digits with a better accuracy, or even over a wider range without worrying about breakage or condensation? Checktemp might just be what you′ve been looking for! It gives you the temperature read-out from -50 to 150°C or -58 to 302°F in seconds on a large Liquid Crystal Display with ±0.3°C or ±0.5°F accuracy. NO breakage, NO waste, NO injuries, NO difficulty in reading. Not even a parallax error by taking a wrong measurement due to the angle of view!

  • Light weight and pocket-sized
  • An NTC thermistor is housed in a stainless steel penetration probe
  • LCD display is encased in a rugged ABS housing
  • Internal calibration device assures the reading is reliable and accurate

Calibration Check Feature

One thing you could say about glass thermometers is that they give a sense of false certainty about the temperature because one can see the physical expansion of the liquid inside the glass! Electronic thermometers have at times been likened to a black box. One is never sure if the display is correct. Well, we′ve solved this problem too. Checktemp has the world′s first cal-check feature. There is a switch under the battery cover which sends a 0.0°C ±0.3°C (32.0°F ±0.5°F) signal to the display. This way you are always sure about the accuracy of your readings and if necessary can manually compensate for any deviation in calibration.

The comparison chart below clearly shows that a Checktemp thermometer is the wisest choice for your routine temperature measurements:

Features:
  • Durability:Checktemp Rugged ABS housing VS Glass/Mercury Made of glass
  • Ease of transport: Checktemp No protection required VS Glass/Mercury Must be protected
  • Response time: Checktemp 20 seconds MAX VS. Glass/Mercury 2 to 5 minutes
  • Easy to read: Checktemp LCD readable at any angle VS. Glass/Mercury Difficult to read
  • Hazardous waste: Checktemp None VS. Glass/Mercury Leaded glass, mercury or alcohol
  • Applications: Checktemp liquid, air, semisolids, frozen material VS. liquid, air

Specifications:

Range:
  • -58.0 to +302°F
  • -50.0 to +150.0°C

Resolution:
  • 0.1°F (-58.0 to 199.9°F); 1°F (200 to 302°F)
  • 0.1°C

Accuracy:
  • ±0.5°F (-4 to +194°F); ±1°F (outside)
  • ±0.3°C (-20 to +90°C); ±0.5°C (outside)

Battery: 1 × 1.4 volt battery
Battery Life: 3000 hours of continuous use
Environment: 32 to 122°F (0 to 50°C); RH 95%
Dimensions: 2.6 × 2.0 × 1 in. (66 × 50 × 25 mm)
Weight: 1.8 oz (50 g)
Probe: Stainless Steel, 105 × 3 mm with a protective cap

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service