Key Documents
Z350419
Sigma-Aldrich
Carestream® BioMax® MR film
Synonym(s):
autoradiography film, xray film
About This Item
UNSPSC Code:
41105341
NACRES:
NB.22
size
14 in. (35 cm) × 17 in. (43 cm)
General description
Each sheet of film is sealed in a light-tight envelope
Single-emulsion film that provides maximum resolution and clearest background. The most sensitive film for direct 14C, 35S, and 33P autoradiography. Higher resolution than X-OMAT AR and BioMax MS for 32P direct and screen exposures and chemiluminescence, but with reduced sensitivity.
Legal Information
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Shujuan Gao et al.
Journal of molecular signaling, 6, 8-8 (2011-08-13)
The family of A-kinase-anchoring proteins, AKAPs, constitutes a group of molecular scaffolds that act to catalyze dynamic interactions of protein kinase A, protein kinase C, tyrosine kinases, G-protein-coupled receptors and ion channels. AKAP5 (MW ~47 kDa) and AKAP12 (MW ~191
Atsuko Kimura et al.
The European journal of neuroscience, 30(2), 299-306 (2009-07-21)
The 5-HT(2C) receptor has been implicated in mood and eating disorders. In general, it is accepted that 5-HT(2C) receptor agonists increase anxiety behaviours and induce hypophagia. However, pharmacological analysis of the roles of these receptors is hampered by the lack
Questions
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?