Key Documents
Z340502
Sigma-Aldrich
Semi-dry blotter
Blotter unit
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
General description
  • Transfer area 20 × 20 cm
  • Low buffer volume
  • Low heat production-Less band distortion
  • Recommended power supplies: Consort EV202 or EV243
  • Includes leads and Mylar shield
Excellent for Western, Northern, or Southern transfers, unit accepts one or several gels up to 20 × 20 cm, and varying thickness.
Stainless steel cathode and platinum anode plates produce highly uniform field and resist corrosion. Mylar shield provided to mask unused area of cathode, prevents arcing, ensures all current passes through gel.
Choose from one of the most recent versions:
Certificates of Analysis (COA)