Key Documents
Z339113
Sigma-Aldrich
Comb for midi horizontal electrophoresis unit
size 1.0 mm × 28 wells
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
grade
electrophoresis grade
feature
well spacing compatible with multichannel pipettes
size
1.0 mm × 28 wells
Choose from one of the most recent versions:
Certificates of Analysis (COA)