Key Documents
Z338834
Sigma-Aldrich
Maxi horizontal electrophoresis units
cooled unit
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
grade
electrophoresis grade
gel W
20 cm
General description
Designed for both analytical and preparative separations of nucleic acids with buffer circulation ports to prevent buffer depletion. Four comb positions permit the screening of up to 160 samples.
Packaging
Includes base unit with hose connectors for circulating coolant, safety lid, one standard casting tray for 20 × 20 cm gels, and two combs, 1.0 mm × 16 sample wells
Choose from one of the most recent versions:
Certificates of Analysis (COA)