Key Documents
Z338818
Sigma-Aldrich
Midi horizontal electrophoresis unit
About This Item
UNSPSC Code:
41105300
grade
electrophoresis grade
gel W
13 cm
General description
Designed for both analytical and preparative separations of nucleic acids. These 13 cm wide units are similar to the Mini units, but with the addition of buffer circulation ports. Four comb positions allow up to 112 samples to be screened on one gel.
Packaging
Includes base unit, safety lid, one standard casting tray for 13 × 15 cm gels, and two combs (1.0 mm × 12 sample wells and 1.0 mm × 16 sample wells).
Choose from one of the most recent versions:
Certificates of Analysis (COA)