Z338796
Mini horizontal electrophoresis unit
complete set
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
grade
electrophoresis grade
gel W
6 cm
General description
Designed for rapid, routine electrophoresis for nucleic acids. The Mini unit is designed with a choice of two casting systems: standard casting trays that require sealing tape; or aluminum-gated casting trays that do not.
Includes base unit, safety lid, two casting trays with aluminum gates (one for 6 × 7 cm and one for 6 × 10 cm gels), and two combs (1.0 mm × 8 sample wells and 1.0 mm × 12 sample wells).
Includes base unit, safety lid, two casting trays with aluminum gates (one for 6 × 7 cm and one for 6 × 10 cm gels), and two combs (1.0 mm × 8 sample wells and 1.0 mm × 12 sample wells).
Choose from one of the most recent versions:
Certificates of Analysis (COA)